Please login to your online account to display your discounted pricing

PBS Tablets, Calbiochem™, EMD Millipore

Dissolving one tablet in 1 liter of deionized H2O yields 140mM NaCl, 10mM phosphate buffer, and 3mM KCl

$57.94 - $57.94

Chemical Identifiers

Chemical Name or Material PBS Tablets
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

5246501EA

 
emd millipore corporation
5246501EA
Each for $57.94
Description & Specifications

Specifications

Quantity 1 strip (10 tablets)
Solubility Information Soluble in water
Recommended Storage 15°C to 30°C (Do not freeze).
Synonym Phosphate Buffered Saline Tablets
Chemical Name or Material PBS Tablets
Safety and Handling

15°C to 30°C (Do not freeze).