Please login to your online account to display your discounted pricing

PBS, Phosphate Buffered Saline, 1X Powder, pH 7.4, Fisher BioReagents


Quantity 10L
Packaging Poly Bottle

Manufacturer: fisher scientific company  FLBP66110

 View more versions of this product

Catalog No. BP661-10

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10L
Protease Protease free
Physical Form Solid
Color White
DNase DNase free
Packaging Poly Bottle
Conductivity 14000 to 17800μmhos/cm
Purity Grade Notes DNase-, RNase- and Protease-Free
CAS 7647-14-5
Wt. of Solid in Pack 98.8 +/- 1.5 or 496 +/- 4g
Chemical Name or Material Phosphate Buffered Saline
CAS 7778-77-0
Name Note 1X Powder Concentrate
ChemAlert Storage Symbol Gray
Synonym PBS
Recommended Storage RT
CAS 7558-79-4
Concentration or Composition (by Analyte or Components) Sodium Chloride (81%), Sodium Phosphate Dibasic (≈14%), Potassium Phosphate Monobasic (≈3.0%), and Potassium Chloride (≈2%).
Chloride 0.136 to 0.144M
CAS 7447-40-7

Specifications

Chemical Name or Material Phosphate Buffered Saline
Name Note 1X Powder Concentrate
CAS 7647-14-5
CAS 7558-79-4
CAS 7778-77-0
CAS 7447-40-7
Purity Grade Notes DNase-, RNase- and Protease-Free
Physical Form Solid
Synonym PBS

  • Provides a convenient way to make 10L of standard 1X phosphate buffer
  • Reconstitute contents of bottle in ultrapure water, such as BP2819, to yield a 1X PBS solution containing 0.137M NaCl, 0.0027M KCl, and 0.0119M phosphates
  • Tested for the absence of DNase, RNase, and protease
  • PBS is a standard buffer used in many biochemical applications to maintain pH
  • Intended for laboratory research use only

Accessories
 

RT

SDS & Safety Documents