missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Pazufloxacin Mesylate, TCI America™

Manufacturer:  TCI America P19621G

 View more versions of this product

Catalog No. P19621G



Specifications

Specifications

Pazufloxacin Mesylate
259°C
163680-77-1
MFCD00913262
UDHGFPATQWQARM-FJXQXJEOSA-N
414.404
414.40
≥98.0% (HPLC,T)
Yellow
1g
C17H19FN2O7S
Pazufloxacin Methanesulfonate
CC1COC2=C3N1C=C(C(=O)C3=CC(=C2C4(CC4)N)F)C(=O)O.CS(=O)(=O)O
6918232
Crystal-Powder at 20°C

Chemical Identifiers

163680-77-1
414.404
UDHGFPATQWQARM-FJXQXJEOSA-N
6918232
C17H19FN2O7S
MFCD00913262
Pazufloxacin Methanesulfonate
CC1COC2=C3N1C=C(C(=O)C3=CC(=C2C4(CC4)N)F)C(=O)O.CS(=O)(=O)O
Safety and Handling

Safety and Handling

RTECSNumber : UU8815400

TSCA : No

Documents