missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Paxilline, 97+%

Potent blocker of high-conductance calcium-activated potassium (BKCa) channels

$242.11 - $956.11

Chemical Identifiers

CAS 57186-25-1
Molecular Formula C27H33NO4
Molecular Weight (g/mol) 435.564
MDL Number MFCD00083464
InChI Key ACNHBCIZLNNLRS-UBGQALKQSA-N
Synonym paxilline, unii-3t9u9z96l7, 2h-1-benzopyrano 5',6':6,7 indeno 1,2-b indol-3 4bh-one, 5,6,6a,7,12,12b,12c,13,14,14a-decahydro-4b-hydroxy-2-1-hydroxy-1-methylethyl-12b,12c-dimethyl-, 2-alpha,4b-beta,6a-alpha,12b-beta,12c-alpha,14a-beta, 2h-pyrano 2,3:5',6' benz 1',2':6,7 indeno 1,2-b indol-3 4bh-one, 5,6,6a,7,12,12b,12c,13,14,14a-decahydro-4b-hydroxy-2-1-hydroxy-1-methylethyl-12b,12c-dimethyl-, 2r,4bs,6as,12bs,12cr,14as, spectrum5_001975, paxilline, powder hplc, paxilline solution, 100 mug/ml in acetonitrile, analytical standard, 2r,4bs,6as,12bs,12cr,14as-5,6,6a,7,12,12b,12c,13,14,14a-decahydro-4b-hydroxy-2-1-hydroxy-1-methylethyl-12b,12c-dimethyl-2h-pyrano 2,3:5',6' benz 1',2':6,7 indeno 1,2-b indol-3 4bh-one, ncgc00025342-07_c27h33no4_ 2r,4bs,6as,12bs,12cr,14as-4b-hydroxy-2-2-hydroxy-2-propanyl-12b,12c-dimethyl-5,6,6a,7,12,12b,12c,13,14,14a-decahydro-2h-chromeno 5',6':6,7 indeno 1,2-b indol-3 4bh-one
PubChem CID 105008
ChEBI CHEBI:34907
SMILES CC12CCC3C(=CC(=O)C(O3)C(C)(C)O)C1(CCC4C2(C5=C(C4)C6=CC=CC=C6N5)C)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ61341LB0 View Documents Alfa Aesar™
J61341LB0
5mg Each for $242.11
 
AAJ61341MB View Documents Alfa Aesar™
J61341MB
25mg Each for $956.11
 
Specifications

Chemical Identifiers

57186-25-1
435.564
ACNHBCIZLNNLRS-UBGQALKQSA-N
105008
CC12CCC3C(=CC(=O)C(O3)C(C)(C)O)C1(CCC4C2(C5=C(C4)C6=CC=CC=C6N5)C)O
C27H33NO4
MFCD00083464
paxilline, unii-3t9u9z96l7, 2h-1-benzopyrano 5',6':6,7 indeno 1,2-b indol-3 4bh-one, 5,6,6a,7,12,12b,12c,13,14,14a-decahydro-4b-hydroxy-2-1-hydroxy-1-methylethyl-12b,12c-dimethyl-, 2-alpha,4b-beta,6a-alpha,12b-beta,12c-alpha,14a-beta, 2h-pyrano 2,3:5',6' benz 1',2':6,7 indeno 1,2-b indol-3 4bh-one, 5,6,6a,7,12,12b,12c,13,14,14a-decahydro-4b-hydroxy-2-1-hydroxy-1-methylethyl-12b,12c-dimethyl-, 2r,4bs,6as,12bs,12cr,14as, spectrum5_001975, paxilline, powder hplc, paxilline solution, 100 mug/ml in acetonitrile, analytical standard, 2r,4bs,6as,12bs,12cr,14as-5,6,6a,7,12,12b,12c,13,14,14a-decahydro-4b-hydroxy-2-1-hydroxy-1-methylethyl-12b,12c-dimethyl-2h-pyrano 2,3:5',6' benz 1',2':6,7 indeno 1,2-b indol-3 4bh-one, ncgc00025342-07_c27h33no4_ 2r,4bs,6as,12bs,12cr,14as-4b-hydroxy-2-2-hydroxy-2-propanyl-12b,12c-dimethyl-5,6,6a,7,12,12b,12c,13,14,14a-decahydro-2h-chromeno 5',6':6,7 indeno 1,2-b indol-3 4bh-one
CHEBI:34907

Specifications

White to Yellow
57186-25-1
MFCD00083464
paxilline, unii-3t9u9z96l7, 2h-1-benzopyrano 5',6':6,7 indeno 1,2-b indol-3 4bh-one, 5,6,6a,7,12,12b,12c,13,14,14a-decahydro-4b-hydroxy-2-1-hydroxy-1-methylethyl-12b,12c-dimethyl-, 2-alpha,4b-beta,6a-alpha,12b-beta,12c-alpha,14a-beta, 2h-pyrano 2,3:5',6' benz 1',2':6,7 indeno 1,2-b indol-3 4bh-one, 5,6,6a,7,12,12b,12c,13,14,14a-decahydro-4b-hydroxy-2-1-hydroxy-1-methylethyl-12b,12c-dimethyl-, 2r,4bs,6as,12bs,12cr,14as, spectrum5_001975, paxilline, powder hplc, paxilline solution, 100 mug/ml in acetonitrile, analytical standard, 2r,4bs,6as,12bs,12cr,14as-5,6,6a,7,12,12b,12c,13,14,14a-decahydro-4b-hydroxy-2-1-hydroxy-1-methylethyl-12b,12c-dimethyl-2h-pyrano 2,3:5',6' benz 1',2':6,7 indeno 1,2-b indol-3 4bh-one, ncgc00025342-07_c27h33no4_ 2r,4bs,6as,12bs,12cr,14as-4b-hydroxy-2-2-hydroxy-2-propanyl-12b,12c-dimethyl-5,6,6a,7,12,12b,12c,13,14,14a-decahydro-2h-chromeno 5',6':6,7 indeno 1,2-b indol-3 4bh-one
ACNHBCIZLNNLRS-UBGQALKQSA-N
435.564
CHEBI:34907
Powder
Paxilline
5mg
C27H33NO4
UN2811
Soluble in DMSO, acetone or chloroform
CC12CCC3C(=CC(=O)C(O3)C(C)(C)O)C1(CCC4C2(C5=C(C4)C6=CC=CC=C6N5)C)O
105008
435.56
≥97%
Safety and Handling

Safety and Handling

GHS H Statement
H300-H310-H330-H318-H315-H335
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement
P260-P301+P310-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

RTECSNumber : DJ2830000

Recommended Storage : −30° to −10°C