Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Paxillin Ab-1, Mouse Monoclonal Antibody

Provide accurate, reproducible results with the Thermo Scientific™ Paxillin Ab-1, Mouse Monoclonal Antibody.

$269.22 - $1,344.84

Specifications

Antigen Paxillin Ab-1
Clone 5H11
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
View More Specs
Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

MS404P

 
thermo scientific™ lab vision™
MS404P
Purified Ab with BSA and Azide 1mL Each for $1,258.12

MS404P0

 
thermo scientific™ lab vision™
MS404P0
Purified Ab with BSA and Azide 100μL Each for $269.22

MS404P1

 
thermo scientific™ lab vision™
MS404P1
Purified Ab with BSA and Azide 500μL Each for $782.31

MS404P1ABX

 
thermo scientific™ lab vision™
MS404P1ABX
Ab without BSA/Azide (Frozen) 100μL Each for $872.61

MS404PABX

 
thermo scientific™ lab vision™
MS404PABX
Ab without BSA/Azide (Frozen) 200μL Each for $1,344.84

MS404R7

 
thermo scientific™ lab vision™
MS404R7
Ready-to-use for IHC (UVLP) 7mL Each for $775.29
Description & Specifications

Specifications

Antigen Paxillin Ab-1
Clone 5H11
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Rat
Cross Reactivity Human
Cross Reactivity Rat
Host Species Murine
Regulatory Status RUO
Research Discipline Cytoskeletal Protein
Type Primary
Immunogen Purified recombinant full-length human paxillin protein
Isotype IgG2a kappa

Provide accurate, reproducible results with the Thermo Scientific™ Paxillin Ab-1, Mouse Monoclonal Antibody.

Paxillin is a focal adhesion protein and a substrate for several tyrosine kinases such as src, FAK, and p210BRC/ABL. The tyrosine phosphorylation of paxillin is affected by conditions that change cell-cell adhesion. Paxillin associates tightly with FAK and Crk through its SH2 domain. This interaction is independent of the extracellular matrix. Although paxillin was initially discovered in fibroblasts, its phosphorylation may also be important during neurite extension during differentiation.

Host Species: Mouse

Clone: 5H11

Isotype: IgG1/κ

Species Reactivity: Human and Rat. Others not known.

Epitope: Not determined

Immunogen: Purified recombinant full-length human paxillin protein

Molecular Weight: 68kDa

Positive Control: HeLa cells, Colon CA

Cellular Localization: Membrane and cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native verified)

Note:

USA: RUO; Int'l: RUO