Parr™ 1341 Plain Jacket Calorimeter
For measuring heat of combustion of solid or liquid fuels, waste materials, and other combustible samples
Manufacturer:  Parr Instrument Co Inc PAR1341EE
 View more versions of this product
Catalog No. 04-350-3
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications
Specifications
21cm
29cm
Ignition unit, molded fiberglass static jacket unit with double walls and a double cover, 6775 digital thermometer, manual oxygen fill, manual bucket fill, manual bomb wash
230V
0.002°C
8.5 in.
21cm
Measuring heat of combustion of solid or liquid fuels, waste materials and other combustible samples
230V 50/60Hz
25 min./test
50/60Hz
5000 to 8000 kcal
ISO 9001:2008, CSA, CE, ASTM
11.5 in.
Manual
2 Tests/Hours
0.003
Plain Jacket Calorimeter
8.5 in.