Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ PARP (Poly ADP-Ribose Polymerase) Ab-3, Rabbit Polyclonal Antibody

Thermo Scientific™ PARP (Poly ADP-Ribose Polymerase) Ab-3, Rabbit Polyclonal Antibody provides accurate, reproducible results.

$277.12 - $1,299.37

Specifications

Antigen PARP (Poly ADP-Ribose Polymerase) Ab-3
Concentration 1mg/mL
Applications Western Blotting
Classification Polyclonal
Conjugate Unlabeled
View More Specs
Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

RB1680P

 
thermo scientific™ lab vision™
RB1680P
Purified Ab with BSA and Azide 1mL Each for $1,218.73

RB1680P0

 
thermo scientific™ lab vision™
RB1680P0
Purified Ab with BSA and Azide 100µL Each for $277.12

RB1680P1

 
thermo scientific™ lab vision™
RB1680P1
Purified Ab with BSA and Azide 500µL Each for $744.39

RB1680P1ABX

 
thermo scientific™ lab vision™
RB1680P1ABX
Ab without BSA/Azide (Frozen) 500µL Each for $833.45

RB1680PABX

 
thermo scientific™ lab vision™
RB1680PABX
Ab without BSA/Azide (Frozen) 1mL Each for $1,299.37
Description & Specifications

Specifications

Antigen PARP (Poly ADP-Ribose Polymerase) Ab-3
Concentration 1mg/mL
Applications Western Blotting
Classification Polyclonal
Conjugate Unlabeled
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Format Concentrated
Host Species Rabbit
Regulatory Status RUO
Research Discipline Apoptosis
Type Primary
Immunogen A synthetic peptide from the C-terminal region of bovine PARP
Isotype IgG

Thermo Scientific™ PARP (Poly ADP-Ribose Polymerase) Ab-3, Rabbit Polyclonal Antibody provides accurate, reproducible results.

Poly ADP-Ribose Polymerase (PARP) uses nicotinamide adenine dinucleotide (oxidized form) NAD as a substrate to catalyse the transfer of ADP-ribose to a variety of nuclear protein acceptors. Proteolysis of PARP to its stable 85kDa fragment is an early marker of programmed cell death (apoptosis) and is mediated by the caspase CPP32 protein. Cleavage occurs between Adp216 and Gly217, a site in PARP conserved across species.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Mouse, Rat and Cow. Others not known.

Epitope: C-terminal

Immunogen: A synthetic peptide from the C-terminal region of bovine PARP

Molecular Weight: 116kDa

Positive Control: Raji cells

Cellular Localization: Nuclear

Recommended for:

  • Western Blotting

Note:

USA: RUO; Int'l: RUO