Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ PARP (Poly ADP-Ribose Polymerase) Ab-3, Rabbit Polyclonal Antibody


Thermo Scientific™ PARP (Poly ADP-Ribose Polymerase) Ab-3, Rabbit Polyclonal Antibody provides accurate, reproducible results.

Manufacturer: thermo scientific™ lab vision™  RB1680P1ABX

 View more versions of this product

Catalog No. RB1680P1ABX

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Antigen PARP (Poly ADP-Ribose Polymerase) Ab-3
Immunogen A synthetic peptide from the C-terminal region of bovine PARP
Host Species Rabbit
Quantity 500µL
Format Concentrated
Applications Western Blotting
Regulatory Status RUO
Isotype IgG
Formulation Ab without BSA/Azide (Frozen)
Research Discipline Apoptosis
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unlabeled
Type Primary
Item Description 500uL; 1mg/mL; Unlabeled; Purified without BSA and azide

Thermo Scientific™ PARP (Poly ADP-Ribose Polymerase) Ab-3, Rabbit Polyclonal Antibody provides accurate, reproducible results.

Poly ADP-Ribose Polymerase (PARP) uses nicotinamide adenine dinucleotide (oxidized form) NAD as a substrate to catalyse the transfer of ADP-ribose to a variety of nuclear protein acceptors. Proteolysis of PARP to its stable 85kDa fragment is an early marker of programmed cell death (apoptosis) and is mediated by the caspase CPP32 protein. Cleavage occurs between Adp216 and Gly217, a site in PARP conserved across species.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Mouse, Rat and Cow. Others not known.

Epitope: C-terminal

Immunogen: A synthetic peptide from the C-terminal region of bovine PARP

Molecular Weight: 116kDa

Positive Control: Raji cells

Cellular Localization: Nuclear

Recommended for:

  • Western Blotting

Note:

USA: RUO; Int'l: RUO