Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ PARP (Poly ADP-Ribose Polymerase) Ab-2, Rabbit Polyclonal Antibody


Deliver accurate, reproducible results with the Thermo Scientific™ PARP (Poly ADP-Ribose Polymerase) Ab-2, Rabbit Polyclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  RB1516P1ABX

 View more versions of this product

Catalog No. RB1516P1ABX

  • / Each
 

Description & Specifications

Specifications

Applications Western Blotting
Immunogen A synthetic peptide for the N-terminal region of human PARP
Isotype IgG
Cross Reactivity Human
Cross Reactivity Mouse
Host Species Rabbit
Regulatory Status RUO
Quantity 500µL
Format Concentrated
Antigen PARP (Poly ADP-Ribose Polymerase) Ab-2
Classification Polyclonal
Research Discipline Apoptosis
Type Primary
Formulation Ab without BSA/Azide (Frozen)
Concentration 1mg/mL
Conjugate Unlabeled

Deliver accurate, reproducible results with the Thermo Scientific™ PARP (Poly ADP-Ribose Polymerase) Ab-2, Rabbit Polyclonal Antibody.

Poly ADP-Ribose Polymerase (PARP) uses nicotinamide adenine dinucleotide (oxidized form) NAD as a substrate to catalyse the transfer of ADP-ribose to a variety of nuclear protein acceptors. Proteolysis of PARP to its stable 85kDa fragment is an early marker of programmed cell death (apoptosis) and is mediated by the caspase CPP32 protein. Cleavage occurs between Adp216 and Gly217, a site in PARP conserved across species.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Mouse and Rat. Others not known.

Epitope: N-terminal

Immunogen: A synthetic peptide for the N-terminal region of human PARP

Molecular Weight: 116kDa

Positive Control: Raji cells, Tonsil

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin only)
  • Western Blotting

Note:

USA: RUO; Int'l: RUO