missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Paromomycin Sulfate, TCI America™

Manufacturer:  TCI America P20921G

 View more versions of this product

Catalog No. P20921G



Specifications

Specifications

Paromomycin Sulfate
C23H47N5O18S
LJRDOKAZOAKLDU-JXDXLELXSA-N
(3S,5S,6S)-5-amino-2-(aminomethyl)-6-[(2R,4S,5S)-5-[(2R,5R,6R)-3,5-diamino-2-[(2S,3S,5S)-3-amino-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-6-hydroxycyclohexyl]oxy-4-hydroxy-2-(hydroxymethyl)oxolan-3-yl]oxyoxane-3,4-diol;sulfuric acid
91972180
≥94.0% (HPLC)
1g
1263-89-4
MFCD00079278
C1C(C(C(C(C1N)OC2C(C(C(C(O2)CO)O)O)N)OC3C(C(C(O3)CO)OC4C(C(C(C(O4)CN)O)O)N)O)O)N.OS(=O)(=O)O
713.706
Crystal-Powder at 20°C
White-Yellow

Chemical Identifiers

1263-89-4
713.706
LJRDOKAZOAKLDU-JXDXLELXSA-N
(3S,5S,6S)-5-amino-2-(aminomethyl)-6-[(2R,4S,5S)-5-[(2R,5R,6R)-3,5-diamino-2-[(2S,3S,5S)-3-amino-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-6-hydroxycyclohexyl]oxy-4-hydroxy-2-(hydroxymethyl)oxolan-3-yl]oxyoxane-3,4-diol;sulfuric acid
C23H47N5O18S
MFCD00079278
91972180
C1C(C(C(C(C1N)OC2C(C(C(C(O2)CO)O)O)N)OC3C(C(C(O3)CO)OC4C(C(C(C(O4)CN)O)O)N)O)O)N.OS(=O)(=O)O
Safety and Handling

Safety and Handling

RTECSNumber : WK2320000

TSCA : No

Documents