Please login to your online account to display your discounted pricing

Alfa Aesar™ Paromomycin sulfate, 50mg/mL in distilled water, sterile-filtered


Quantity 25mL

Manufacturer: alfa aesar™  J67447AC

 View more versions of this product

Catalog No. AAJ67447AC

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material Paromomycin sulfate
MDL Number MFCD00079278
CAS 1263-89-4
Name Note 50mg/mL in distilled water, sterile-filtered
RTECS Number WK2320000
Physical Form Liquid
Quantity 25mL
Storage Note 1 -30° to -10°C

Specifications

Chemical Name or Material Paromomycin sulfate
CAS 1263-89-4
Name Note 50mg/mL in distilled water, sterile-filtered
MDL Number MFCD00079278
Accessories

-30° to -10°C