Please login to your online account to display your discounted pricing

Alfa Aesar™ Paromomycin sulfate, 50mg/mL in distilled water, sterile-filtered

Chemical Identifiers

Chemical Name or Material Paromomycin sulfate
CAS 1263-89-4
Name Note 50mg/mL in distilled water, sterile-filtered
MDL Number MFCD00079278
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67447AD

 
alfa aesar™
J67447AD
50mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67447AC

 
alfa aesar™
J67447AC
25mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Name Note 50mg/mL in distilled water, sterile-filtered
MDL Number MFCD00079278
CAS 1263-89-4
Chemical Name or Material Paromomycin sulfate
Physical Form Liquid
Storage Note 1 -30° to -10°C
Quantity 50mL
RTECS Number WK2320000
Safety and Handling

-30° to -10°C