Please login to your online account to display your discounted pricing
Parker domnick hunter™ Nitrogen Generators for GC
Specifications
For Use With (Application) GC and other critical analytical applications
Nitrogen Purity >99.9995%
View More Specs
Products
Catalog Number Mfr. No. Electrical Requirements Flow Rate Includes Price Quantity    
14-950-613
parker domnick hunter™
UHPN2-550C-W
103-126V 60Hz 0.55L/min With Integral Compressor CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications
Specifications
For Use With (Application) GC and other critical analytical applications
Nitrogen Purity >99.9995%