Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Parathyroid Hormone Receptor (PTH-Receptor) Type 1 Ab-1, Mouse Monoclonal Antibody

Deliver accurate, reproducible results with the Thermo Scientific™ Parathyroid Hormone Receptor (PTH-Receptor) Type 1 Ab-1, Mouse Monoclonal Antibody.

$305.73 - $1,407.20

Specifications

Antigen Parathyroid Hormone Receptor (PTH-Receptor) Type 1
Clone 3D1.1
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Classification Monoclonal
View More Specs
Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

MS1270P

 
thermo scientific
MS1270P
Purified Ab with BSA and Azide 1mL Each for $1,283.18

MS1270P0

 
thermo scientific™ lab vision™
MS1270P0
Purified Ab with BSA and Azide 100µL Each for $305.73

MS1270P1

 
thermo scientific™ lab vision™
MS1270P1
Purified Ab with BSA and Azide 500µL Each for $767.08

MS1270P1ABX

 
thermo scientific
MS1270P1ABX
Ab without BSA/Azide (Frozen) 100µL Each for $864.08

MS1270PABX

 
thermo scientific™ lab vision™
MS1270PABX
Ab without BSA/Azide (Frozen) 200µL Each for $1,407.20

MS1270R7

 
thermo scientific
MS1270R7
Ready-to-use for IHC (UVLP) 7mL Each for $672.24
Description & Specifications

Specifications

Antigen Parathyroid Hormone Receptor (PTH-Receptor) Type 1
Clone 3D1.1
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Host Species Mouse
Regulatory Status RUO
Research Discipline Steroids Hormones and Growth Factors
Type Primary
Immunogen A 23 amino acid peptide of the hPTH/PTHrP receptor which is not shared by the type II receptor
Isotype IgG2a kappa

Deliver accurate, reproducible results with the Thermo Scientific™ Parathyroid Hormone Receptor (PTH-Receptor) Type 1 Ab-1, Mouse Monoclonal Antibody.

Two human parathyroid hormone (hPTH) receptors are known: one that binds both hPTH and a PTH-related protein called PTHrP (the Type I receptor); and one that only binds hPTH (the Type II receptor).

Host Species: Mouse

Clone: 3D1.1

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: aa 146-169

Immunogen: A 23 amino acid peptide of the hPTH/PTHrP receptor which is not shared by the type II receptor

Molecular Weight: 80 and 160kDa

Positive Control: Breast CA

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting

Note:

USA: RUO; Int'l: RUO