Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Parathyroid Hormone Receptor (PTH-Receptor) Type 1 Ab-1, Mouse Monoclonal Antibody


Deliver accurate, reproducible results with the Thermo Scientific™ Parathyroid Hormone Receptor (PTH-Receptor) Type 1 Ab-1, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  MS1270PABX

 View more versions of this product

Catalog No. MS1270PABX

  • / Each
 

Description & Specifications

Specifications

Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Isotype IgG2a kappa
Quantity 200µL
Host Species Mouse
Format Concentrated
Clone 3D1.1
Regulatory Status RUO
Antigen Parathyroid Hormone Receptor (PTH-Receptor) Type 1
Immunogen A 23 amino acid peptide of the hPTH/PTHrP receptor which is not shared by the type II receptor
Cross Reactivity Human
Formulation Ab without BSA/Azide (Frozen)
Type Primary
Conjugate Unlabeled
Research Discipline Steroids Hormones and Growth Factors
Classification Monoclonal
Concentration 1mg/mL

Deliver accurate, reproducible results with the Thermo Scientific™ Parathyroid Hormone Receptor (PTH-Receptor) Type 1 Ab-1, Mouse Monoclonal Antibody.

Two human parathyroid hormone (hPTH) receptors are known: one that binds both hPTH and a PTH-related protein called PTHrP (the Type I receptor); and one that only binds hPTH (the Type II receptor).

Host Species: Mouse

Clone: 3D1.1

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: aa 146-169

Immunogen: A 23 amino acid peptide of the hPTH/PTHrP receptor which is not shared by the type II receptor

Molecular Weight: 80 and 160kDa

Positive Control: Breast CA

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting

Note:

USA: RUO; Int'l: RUO