missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Pararosaniline 97.0+%, TCI America™

Manufacturer:  TCI America P0021500G

 View more versions of this product

Catalog No. P0021500G



Specifications

Specifications

Pararosaniline
205°C
467-62-9
MFCD00036222
KRVRUAYUNOQMOV-UHFFFAOYSA-N
tris(4-aminophenyl)methanol
10084
Crystal-Powder at 20°C
Purple-Red
500g
C19H19N3O
pararosaniline base, tris 4-aminophenyl methanol, tris p-aminophenyl methanol, benzenemethanol, 4-amino-.alpha.,.alpha.-bis 4-aminophenyl, tris 4-aminophenyl methan-1-ol, fuchsin dye base, parafuchsin carbinol, tris 4-aminophenyl carbinol, methanol, tris 4-aminophenyl, 4,4',4-triaminotrityl alcohol
C1=CC(=CC=C1C(C2=CC=C(C=C2)N)(C3=CC=C(C=C3)N)O)N
305.381
305.38
≥97.0% (T)

Chemical Identifiers

467-62-9
305.381
KRVRUAYUNOQMOV-UHFFFAOYSA-N
10084
C1=CC(=CC=C1C(C2=CC=C(C=C2)N)(C3=CC=C(C=C3)N)O)N
C19H19N3O
MFCD00036222
pararosaniline base, tris 4-aminophenyl methanol, tris p-aminophenyl methanol, benzenemethanol, 4-amino-.alpha.,.alpha.-bis 4-aminophenyl, tris 4-aminophenyl methan-1-ol, fuchsin dye base, parafuchsin carbinol, tris 4-aminophenyl carbinol, methanol, tris 4-aminophenyl, 4,4',4-triaminotrityl alcohol
tris(4-aminophenyl)methanol
Safety and Handling

Safety and Handling

TSCA : Yes

Documents