missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Pararosaniline base, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 201550100
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Pararosaniline base | |
205.0°C | |
Authentic | |
C19H19N3O | |
10 g | |
pararosaniline base, tris 4-aminophenyl methanol, tris p-aminophenyl methanol, benzenemethanol, 4-amino-.alpha.,.alpha.-bis 4-aminophenyl, tris 4-aminophenyl methan-1-ol, fuchsin dye base, parafuchsin carbinol, tris 4-aminophenyl carbinol, methanol, tris 4-aminophenyl, 4,4',4-triaminotrityl alcohol | |
KRVRUAYUNOQMOV-UHFFFAOYSA-N | |
tris(4-aminophenyl)methanol | |
10084 | |
Crystalline Powder |
467-62-9 | |
Purple | |
Glass bottle | |
MFCD00036222 | |
13, 733 | |
Solubility in water: very slightly soluble. Other solubilities: soluble in alcohol,slightly soluble in ether | |
C1=CC(=CC=C1C(C2=CC=C(C=C2)N)(C3=CC=C(C=C3)N)O)N | |
305.38 | |
305.38 |
Chemical Identifiers
467-62-9 | |
305.38 | |
KRVRUAYUNOQMOV-UHFFFAOYSA-N | |
10084 | |
C1=CC(=CC=C1C(C2=CC=C(C=C2)N)(C3=CC=C(C=C3)N)O)N |
C19H19N3O | |
MFCD00036222 | |
pararosaniline base, tris 4-aminophenyl methanol, tris p-aminophenyl methanol, benzenemethanol, 4-amino-.alpha.,.alpha.-bis 4-aminophenyl, tris 4-aminophenyl methan-1-ol, fuchsin dye base, parafuchsin carbinol, tris 4-aminophenyl carbinol, methanol, tris 4-aminophenyl, 4,4',4-triaminotrityl alcohol | |
tris(4-aminophenyl)methanol |
Safety and Handling
GHS H Statement
Suspected of causing cancer.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
GHS Signal Word: Warning
EINECSNumber : 207-395-
RUO – Research Use Only