missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific™ Paraformaldehyde Solution, 4% in PBS, Thermo Scientific™
Manufacturer: Thermo Scientific™ J19943K2
Specifications
Paraformaldehyde Solution | |
Plastic bottle | |
MFCD00133991 |
4% in PBS | |
1L | |
Solution |
Safety and Handling
P201-P202-P261-P272-P280g-P281-P302+P352-P305+P351+P338-P308+P313-P310-P333+P313-P363-P501c
H317-H318-H351
TSCA : Yes
Recommended Storage : Keep cold
Promotions
-
Fisher Chemical
,
Fisher BioReagents
Get Up to $1,500 in Chemicals with Purchase
Spend up to $2,000 on Fisher Chemical, Fisher Bioreagents, Acros Organics, and Alfa Aesar products to earn.: 06/30/21
-
Fisher BioReagents
Get 2 Extra Life Sciences Chemicals When You Buy 3
Buy three life sciences chemicals and get two more at no additional cost.: 03/31/21
-
Thermo Scientific
,
Fisher Chemical
,
Fisher BioReagents
Spend More, Get More on Chemicals
Get up to $1,800 in select chemicals when you spend $1,200 or more.: 06/30/21
-
Get a Molecular Model Set with $250 Purchase
Spend $250 or more on select chemicals and get a molecular model set at no additional cost.: 03/31/21