Please login to your online account to display your discounted pricing

Paraformaldehyde, BAKER™, J.T.Baker™


A grade of chemicals for laboratory use

Quantity 3kg
Packaging Poly Pail

Manufacturer: avantor performance  S89809

 View more versions of this product

Catalog No. 02003575

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 3kg
Chemical Name or Material Paraformaldehyde
CAS 30525-89-4
Molecular Formula (CH2O)n
Specific Gravity 1L = 1.45kg
Packaging Poly Pail
Flash Point 70.00°C

Specifications

Chemical Name or Material Paraformaldehyde
CAS 30525-89-4
Molecular Formula (CH2O)n