Please login to your online account to display your discounted pricing

Paraformaldehyde, BAKER™, J.T.Baker™

A grade of chemicals for laboratory use

$123.77 - $299.85

Chemical Identifiers

Chemical Name or Material Paraformaldehyde
CAS 30525-89-4
Molecular Formula (CH2O)n
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003575

 
avantor performance
S89809
Poly Pail 3kg Each for $299.85

02003576

 
avantor performance
S89807
Poly Bottle 500g Each for $123.77
Description & Specifications

Specifications

Quantity 3kg
Chemical Name or Material Paraformaldehyde
CAS 30525-89-4
Molecular Formula (CH2O)n
Specific Gravity 1L = 1.45kg
Packaging Poly Pail
Flash Point 70.00°C