missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Paraformaldehyde, 97%

Manufacturer:  Alfa Aesar™ A1131336

 View more versions of this product

Catalog No. AAA1131336



Specifications

Specifications

Paraformaldehyde
CH2O
UN2213
14,7025
WSFSSNUMVMOOMR-UHFFFAOYSA-N
formaldehyde
712
97%
500g
30525-89-4
MFCD00133991
102769
formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde
C=O
30.026
CHEBI:16842
120°C to 170°C (decomposition)
71°C (159°F)

Chemical Identifiers

30525-89-4
30.026
WSFSSNUMVMOOMR-UHFFFAOYSA-N
712
formaldehyde
CH2O
MFCD00133991
formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde
CHEBI:16842
C=O
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H228-H351-H302-H332-H317
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable solid.
Suspected of causing cancer.
Harmful if swallowed.
Harmful if inhaled.
May cause an allergic skin reaction.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: PARAFORMALDEHYDE

RTECSNumber : RV0540000

TSCA : TSCA

SDS Documents