Learn More
Paraformaldehyde, 90%, pure, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 169650025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Paraformaldehyde | |
120.0°C to 170.0°C | |
3.5 to 5 (10% suspension) | |
Authentic | |
Plastic bottle | |
(CH2O)n | |
01,566 | |
formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde | |
WSFSSNUMVMOOMR-UHFFFAOYSA-N | |
formaldehyde | |
712 | |
90% | |
Crystalline Powder or Crystals |
30525-89-4 | |
White to off-white | |
71°C | |
89.5 to 93% | |
CH2O | |
2.5 kg | |
15,7129 | |
Solubility in water: sparingly soluble. Other solubilities: the higher polymers are insoluble in water, insoluble in alcohol and ether | |
C=O | |
30.026 | |
CHEBI:16842 | |
Pure |
Chemical Identifiers
30525-89-4 | |
30.026 | |
formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde | |
CHEBI:16842 | |
C=O |
CH2O | |
WSFSSNUMVMOOMR-UHFFFAOYSA-N | |
712 | |
formaldehyde |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye damage.
Suspected of causing cancer.
May cause an allergic skin reaction.
Harmful if inhaled.<br
GHS P Statement
Wear eye protection/face protection.
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF
GHS Signal Word: Danger
RUO – Research Use Only