missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Paraformaldehyde, 16% w/v aq. soln., methanol free, Alfa Aesar™

Manufacturer:  Alfa Aesar 433689M

 View more versions of this product

Catalog No. AA433689M



Description

Description

  • Ampouled under argon
Specifications

Specifications

Paraformaldehyde
Micro filtered
CH2O
14,7025
WSFSSNUMVMOOMR-UHFFFAOYSA-N
formaldehyde
712
Liquid, Ampouled Under Argon
10 x 10mL
30525-89-4
MFCD00133991
formalin
C=O
30.026
CHEBI:16842

Chemical Identifiers

30525-89-4
30.026
WSFSSNUMVMOOMR-UHFFFAOYSA-N
712
formaldehyde
CH2O
MFCD00133991
formalin
CHEBI:16842
C=O
Safety and Handling

Safety and Handling

GHS H Statement
H351-H315-H319-H317

GHS P Statement
P201-P261-P280-P281-P305+P351+P338-P362-P363-P308+P313-P405-P501a

Warning

TSCA : TSCA

storageNote1 :

  • Ampouled under argon

Documents