missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Paraffin wax, pure, granular, ACROS Organics™

Paraffin wax, CnH2n+2, CAS Number-8002-74-2, propafenona inn-spanish, propafenona, propafenone-hcl, propafenonum inn-latin, propafenonum, propafenone inn:ban, propafenone inn, rythmol, propafenone, gnf-pf-4594, 2kg, 322.0 deg.C, CHEBI:63619, White, 232-315-6, 198 deg.C, Pure

Supplier:  Thermo Scientific Chemicals 416770020

 View more versions of this product

Catalog No. AC416770020


Description

Description

Paraffin wax, CnH2n+2, CAS Number-8002-74-2, propafenona inn-spanish, propafenona, propafenone-hcl, propafenonum inn-latin, propafenonum, propafenone inn:ban, propafenone inn, rythmol, propafenone, gnf-pf-4594, 2kg, 322.0 deg.C, CHEBI:63619, White, 232-315-6, 198 deg.C, Pure

Specifications

Specifications

Paraffin wax
58.0°C to 62.0°C
322.0°C
Authentic
CnH2n+2
2 kg
propafenone, rythmol, propafenona, propafenonum, propafenonum inn-latin, propafenona inn-spanish, propafenone inn:ban, gnf-pf-4594, propafenone-hcl, propafenone inn
JWHAUXFOSRPERK-UHFFFAOYSA-N
1-[2-[2-hydroxy-3-(propylamino)propoxy]phenyl]-3-phenylpropan-1-one
4932
CHEBI:63619
Pellets
8002-74-2
White
198°C
Plastic bottle
MFCD00132833
14,7023
Solubility in water: insoluble. Other solubilities: insoluble in alcohol (soluble in warm alcohol),insoluble in acids,soluble in benzene,chloroform,ether,carbon,disulfide,ligroin,turpentine,miscible when melted with wax,fats
CCCNCC(COC1=CC=CC=C1C(=O)CCC2=CC=CC=C2)O
341.451
3-4.5 mPa.s (98.9°C)
Pure

Chemical Identifiers

8002-74-2
341.451
JWHAUXFOSRPERK-UHFFFAOYSA-N
4932
1-[2-[2-hydroxy-3-(propylamino)propoxy]phenyl]-3-phenylpropan-1-one
CnH2n+2
MFCD00132833
propafenone, rythmol, propafenona, propafenonum, propafenonum inn-latin, propafenona inn-spanish, propafenone inn:ban, gnf-pf-4594, propafenone-hcl, propafenone inn
CHEBI:63619
CCCNCC(COC1=CC=CC=C1C(=O)CCC2=CC=CC=C2)O
Safety and Handling

Safety and Handling

EINECSNumber : 232-315-6

RTECSNumber : RV0350000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only