Please login to your online account to display your discounted pricing

Paraffin Oil, BAKER™, J.T.Baker™


A grade of chemicals for laboratory use

Quantity 4L
Packaging Narrow Mouth Amber Glass Bottle

Manufacturer: avantor performance  S89400

 View more versions of this product

Catalog No. 02003572

  • / Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material Paraffin Oil
CAS 8012-95-1
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 0.88kg
Quantity 4L
Flash Point 135.00°C

Specifications

Chemical Name or Material Paraffin Oil
CAS 8012-95-1