missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Paraffin, liquid, pure, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 171402500

 View more versions of this product

Catalog No. AC171402500



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Paraffin, liquid
8012-95-1
Colorless
300.0°C
Authentic
MFCD00131611
250 mL
14, 7186
Solubility in water: insoluble
*
0.00
65-75 mm2/s (40°C)
Pure
Pure
-24.0°C
0.8800g/mL
218°C
Glass bottle
MFCD00131611
0.88
delphinidin chloride, delphinidin, delphinidine, delphinidol, ephdine, delfinidol chloride, unii-em6md4aehe, ccris 2518, em6md4aehe, 3,5,7-trihydroxy-2-3,4,5-trihydroxyphenyl benzopyrylium chloride
FFNDMZIBVDSQFI-UHFFFAOYSA-N
2-(3,4,5-trihydroxyphenyl)chromenylium-3,5,7-triol;chloride
68245
CHEBI:38701
Oily Liquid

Chemical Identifiers

8012-95-1
0.00
FFNDMZIBVDSQFI-UHFFFAOYSA-N
68245
2-(3,4,5-trihydroxyphenyl)chromenylium-3,5,7-triol;chloride
MFCD00131611
MFCD00131611
delphinidin chloride, delphinidin, delphinidine, delphinidol, ephdine, delfinidol chloride, unii-em6md4aehe, ccris 2518, em6md4aehe, 3,5,7-trihydroxy-2-3,4,5-trihydroxyphenyl benzopyrylium chloride
CHEBI:38701
*
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 232-384-2

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only