missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Parabanic Acid 98.0+%, TCI America™

Manufacturer:  TCI America P01951G

 View more versions of this product

Catalog No. P01951G



Specifications

Specifications

Parabanic Acid
249°C
120-89-8
MFCD00014493
ZFLIKDUSUDBGCD-UHFFFAOYSA-N
imidazolidine-2,4,5-trione
67126
114.06
≥98.0% (T)
White
1g
C3H2N2O3
N,N′C-Oxalylurea
C1(=O)C(=O)NC(=O)N1
114.06
CHEBI:74661
Crystal-Powder at 20°C

Chemical Identifiers

120-89-8
114.06
ZFLIKDUSUDBGCD-UHFFFAOYSA-N
67126
imidazolidine-2,4,5-trione
C3H2N2O3
MFCD00014493
N,N′C-Oxalylurea
CHEBI:74661
C1(=O)C(=O)NC(=O)N1
Safety and Handling

Safety and Handling

TSCA : Yes

Documents