Please login to your online account to display your discounted pricing

Alfa Aesar™ Papaverine hydrochloride, 99%

$47.16 - $47.16

Chemical Identifiers

Chemical Name or Material Papaverine hydrochloride
CAS 61-25-6
Molecular Formula C20H21NO4·HCl
Formula Weight 375.85
MDL Number MFCD00012745
Merck Index 14,7019
Synonym 1-(3,4-Dimethoxybenzyl)-6,7-dimethoxyisoquinoline hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2541206

  SDS
alfa aesar™
B2541206
5g Each for $47.16
Description & Specifications

Specifications

Formula Weight 375.85
Chemical Name or Material Papaverine hydrochloride
TSCA TSCA
Assay 99%
Molecular Formula C20H21NO4·HCl
MDL Number MFCD00012745
CAS 61-25-6
Merck Index 14,7019
Synonym 1-(3,4-Dimethoxybenzyl)-6,7-dimethoxyisoquinoline hydrochloride
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302
Harmful if swallowed.
Melting Point ≈220°C (dec.)
Quantity 5g
Health Hazard 3 GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number NW8575000

Solubility
Soluble in water (50 mg/ml), DMSO, methanol, 100% ethanol and chloroform.

Notes
Light sensitive. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302
Harmful if swallowed.