missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Papanicolaou Stain, EA-36-50 Formula, Counterstain for use with Hematoxylin Stains in the Papanicolaou Staining Procedure, Ricca Chemical

$140.58 - $140.58

Chemical Identifiers

CAS 64-17-5,7732-18-5,17372-87-1,12067-99-1,10114-58-6,554-13-2
CAS Min % 90.0,6.0,0.2,0.23,0.06,9.0E-4
CAS Max % 91.0,8.0,0.3,0.24,0.06,9.0E-4
Physical Form Murky Liquid
Chemical Name or Material Papanicolaou Stain, EA-36-50 Formula
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
5500-16 SDS Ricca Chemical Company
RI550016
Poly natural 500mL Each for $140.58
5500-1 SDS Ricca Chemical Company
RI55001
Poly natural 4L N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
5500-32 Ricca Chemical Company
RI550032
Poly natural 1L N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

Counterstain For use with Hematoxylin Stains in the Papanicolaou Staining Procedure
Description & Specifications

Chemical Identifiers

64-17-5,7732-18-5,17372-87-1,12067-99-1,10114-58-6,554-13-2
91.0,8.0,0.3,0.24,0.06,9.0E-4
Papanicolaou Stain, EA-36-50 Formula
90.0,6.0,0.2,0.23,0.06,9.0E-4
Murky Liquid

Specifications

Murky Liquid
500mL
9.0E-4,0.2,90.0,6.0,0.06,0.23
Poly natural
Green
64-17-5,7732-18-5,554-13-2,17372-87-1,10114-58-6,12067-99-1
0.06,9.0E-4,8.0,0.24,0.3,91.0
Papanicolaou Stain, EA-36-50 Formula