Please login to your online account to display your discounted pricing

Pancuronium dibromide, Tocris


Nicotinic (neuromuscular) antagonist

Quantity 50mg

Manufacturer: r & d systems  RDS069350

 View more versions of this product

Catalog No. 069350

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 50mg
Formula Weight 732.68
Assay Percent Range >99%
CAS 15500-66-0
Molecular Formula C35H60Br2N2O4
Chemical Name or Material Pancuronium dibromide
Synonym 1,1'-(3α,17β-Dihydroxy-2β,5α-androstan-2β,16β-ylene) bis[1-methylpiperidinium] diacetate dibromide
Recommended Storage Desiccate at Room Temperature

Specifications

Chemical Name or Material Pancuronium dibromide
Assay Percent Range >99%
Molecular Formula C35H60Br2N2O4
Formula Weight 732.68
Synonym 1,1'-(3α,17β-Dihydroxy-2β,5α-androstan-2β,16β-ylene) bis[1-methylpiperidinium] diacetate dibromide
Accessories

Desiccate at Room Temperature