Please login to your online account to display your discounted pricing

Alfa Aesar™ Pancreastatin (33-49), Porcine

For cellular and molecular biology applications

Specifications

Product Type Pancreastatin (33-49), porcine
Cross Reactivity Porcine
Molecular Weight 1846.93
Storage Requirements -30° to -10°C
Sequence Gln-Glu-Glu-Glu-Glu-Glu-Thr-Ala-Gly-Ala-Pro-Gln-Gly-Leu-Phe-Arg-Gly-NH2
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66504EXD

 
alfa aesar™
J66504EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66504MCR

 
alfa aesar™
J66504MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Pancreastatin (33-49), porcine
Cross Reactivity Porcine
Molecular Weight 1846.93
Storage Requirements -30° to -10°C
Sequence Gln-Glu-Glu-Glu-Glu-Glu-Thr-Ala-Gly-Ala-Pro-Gln-Gly-Leu-Phe-Arg-Gly-NH2
Format Solid
Regulatory Status RUO
Purification Method HPLC