Please login to your online account to display your discounted pricing

PAN Indicator, 0.3% (w/v) in Ethanol, Ricca Chemical

$66.91 - $66.91

Chemical Identifiers

Chemical Name or Material PAN Indicator
CAS 64-17-5
CAS Min % 88.0
CAS Max % 92.0
CAS 67-63-0
CAS Min % 4.0
CAS Max % 6.0
CAS 67-56-1
CAS Min % 4.0
CAS Max % 6.0
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

5487-34

 
ricca chemical company
RI5487.34
Glass amber 120mL Each for $66.91
Description & Specifications

Specifications

Physical Form Liquid
Color Clear amber
Quantity 120mL
Chemical Name or Material PAN Indicator
Concentration or Composition (by Analyte or Components) 0.3% (w/v) in Ethanol
Packaging Glass amber
CAS 64-17-5
CAS 67-56-1
CAS 67-63-0
CAS Min % 88.0
CAS 85-85-8
CAS Min % 0.37
CAS Min % 4.0
CAS Max % 92.0
CAS Max % 6.0
CAS Max % 6.0
CAS Min % 4.0
CAS Max % 0.38
Shelf Life 24 months
Safety and Handling

24 months