Learn More
Pan Cytokeratin Monoclonal Antibody (AE1/AE3), eFluor™ 615, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$640.00 - $640.00
Specifications
Antigen | Pan Cytokeratin |
---|---|
Clone | AE1/AE3 |
Concentration | 0.2 mg/mL |
Classification | Monoclonal |
Conjugate | eFluor 615 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
50245828
|
Invitrogen
42900382 |
100 μg |
Each for $640.00
|
|
|||||
Description
Description: The monoclonal antibodies AE1 and AE3 recognize many of the acidic and basic cytokeratin family members. Cytokeratins are intermediate filament proteins comprising one component of the cytoskeleton. There are two large families of cytokeratins, acidic and basic, but all contain the same basic domains (i.e. an alpha-helical core with an N- and C-terminal domain). The proteins are expressed in epithelial cells, but are developmentally regulated. Many tumors also express these proteins and their expression can help identify the origin of a neoplasm. The AE3 monoclonal antibody recognizes the 65 to 67 triplet, 64, 59, 58, 56, 54 and 52kD proteins also known as cytokeratin 1, 2, 3, 4, 5, 6, 7, and 8 while the AE1 antibody recognizes 56.5, 54', 50, 50', 48, and 40 kDa proteins (also known as CK10, 14, 15, 16 and 19). These antibodies can be used on a wide array of tissue samples from mouse, human, rat, primates (cynomolgus and rhesus), dog, cat, rabbit, and chicken. Applications Reported: This AE1/AE3 antibody has been reported for use in immunohistochemical staining, immunocytochemistry, immunohistochemical staining of frozen tissue sections, and immunohistochemical staining of formalin-fixed paraffin embedded tissue sections. Applications Tested: This AE1/AE3 antibody has been tested by immunocytochemistry on methanol-fixed MCF7 cells and can be used at less than or equal to 5 μg/mL.
Pan Cytokeratin Monoclonal antibody specifically detects Pan Cytokeratin in Canine, Human, Mouse, Non-human Primate, Rabbit, Rhesus Monkey samples. It is validated for Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin).Specifications
Pan Cytokeratin | |
0.2 mg/mL | |
eFluor 615 | |
Mouse | |
Canine, Human, Mouse, Non-human Primate, Rabbit, Rhesus Monkey | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
KRT14 |
AE1/AE3 | |
Monoclonal | |
Liquid | |
RUO | |
IgG1 | |
Affinity chromatography | |
Antibody |
For Research Use Only.