missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Pan Cytokeratin (AE1/AE3) Mouse, eFluor 570, Clone: AE1/AE3, eBioscience™

Mouse Monoclonal Antibody

$146.52 - $361.68

Specifications

Antigen Pan Cytokeratin (AE1/AE3)
Clone AE1/AE3
Host Species Murine
Gene Alias keratin, basic, acidic
Applications Immunocytochemistry,Immunohistochemistry,Immunohistochemistry (Formalin/Paraffin),Immunohistochemistry (Frozen)
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501123640 Life Technologies
LS41900380
25μg Each for $146.52
501123641 Life Technologies
LS41900382
100μg Each for $361.68
Specifications

Description

The monoclonal antibodies AE1 and AE3 recognize many of the acidic and basic cytokeratin family members. Cytokeratins are intermediate filament proteins comprising one component of the cytoskeleton. There are two large families of cytokeratins, acidic and basic, but all contain the same basic domains (i.e. an alpha-helical core with an N- and C-terminal domain). The proteins are expressed in epithelial cells, but are developmentally regulated. Many tumors also express these proteins and their expression can help identify the origin of a neoplasm.

The AE3 monoclonal antibody recognizes the 65 to 67 triplet, 64, 59, 58, 56, 54 and 52kD proteins also known as cytokeratin 1, 2, 3, 4, 5, 6, 7, and 8 while the AE1 antibody recognizes 56.5, 54’, 50, 50’, 48, and 40kDa proteins (also known as CK10, 14, 15, 16 and 19). These antibodies can be used on a wide array of tissue samples from mouse, human, rat, primates (cynomolgus and rhesus), dog, cat, rabbit, and chicken.
Description & Specifications

Specifications

Pan Cytokeratin (AE1/AE3)
Murine
Immunocytochemistry,Immunohistochemistry,Immunohistochemistry (Formalin/Paraffin),Immunohistochemistry (Frozen)
eFluor 570
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL
AE1/AE3
keratin, basic, acidic
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer