Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Cadherin, Pan Ab-4, Rabbit Polyclonal Antibody


Ensure accurate, reproducible results in immunohistochemistry experiments with Thermo Scientific™ Cadherin, Pan Ab-4, Rabbit Polyclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  RB1524P1ABX

 View more versions of this product

Catalog No. RB1524P1ABX

  • / Each
 

Description & Specifications

Specifications

Regulatory Status RUO
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Chicken
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Non-Human Primate
Cross Reactivity Rat
Format Concentrated
Quantity 500µL
Antigen Cadherin, Pan Ab-4
Isotype IgG
Host Species Rabbit
Applications Immunohistochemistry (Paraffin)
Immunogen A synthetic peptide from the C-terminal end of chicken N-cadherin
Formulation Ab without BSA/Azide (Frozen)
Research Discipline Junction Protein
Type Primary
Classification Polyclonal
Purification Method Tonsil, squamous epithelium
Concentration 1mg/mL
Conjugate Unlabeled
Item Description 500uL; 1mg/mL; Purified w/o BSA, azide

Ensure accurate, reproducible results in immunohistochemistry experiments with Thermo Scientific™ Cadherin, Pan Ab-4, Rabbit Polyclonal Antibody.

Cadherins are calcium-dependent cell adhesion molecules, which play important role in the growth and development of cells via the mechanisms of control of tissue architecture and the maintenance of tissue integrity. They are expressed in a tissue-specific manner and are required for assembly of cells into solid tissue. Individual cadherin molecules are known to cooperate with each other to form a linear cell adhesion zipper. In adhesion junctions cadherins are bound to Beta and gamma catenins which in turn bind to alpha catenin, an actin binding protein. Cadherins play important part in tumor invasion and metastasis.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Mouse, Rat, Chicken, Dog, Monkey, Cow and Xenopus.

Epitope: C-terminal

Immunogen: A synthetic peptide from the C-terminal end of chicken N-cadherin

Positive Control: Tonsil, squamous epithelium

Cellular Localization: Cell membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO