Please login to your online account to display your discounted pricing

PAN, BAKER ANALYZED™ Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$137.70 - $311.76

Chemical Identifiers

Chemical Name or Material PAN
CAS 85-85-8
Purity Grade BAKER ANALYZED™ Reagent
Molecular Formula C5H4NN:NC10H6OH
Formula Weight 249.28
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003570

 
avantor performance
S89100
Wide Mouth Amber Glass Bottle 1g Each for $137.70

02003571

 
avantor performance
S89101
Wide Mouth Amber Glass Bottle 5g Each for $311.76
Description & Specifications

Specifications

Formula Weight 249.28
Chemical Name or Material PAN
Purity Grade BAKER ANALYZED™ Reagent
Molecular Formula C5H4NN:NC10H6OH
Packaging Wide Mouth Amber Glass Bottle
CAS 85-85-8
Quantity 1g