Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Actin, pan Ab-5, Mouse Monoclonal Antibody


Obtain accurate, reproducible results in immunohistochemistry, western blotting, and immunoprecipitation experiments.

Manufacturer: thermo scientific™ lab vision™  MS1295PABX

 View more versions of this product

Catalog No. MS1295PABX

  • / Each
 

Description & Specifications

Specifications

Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Host Species Mouse
Immunogen Chicken gizzard actin
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Chicken
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Physarum polycephalum
Cross Reactivity Porcine
Cross Reactivity Rabbit
Cross Reactivity Rat
Quantity 200µL
Antigen Actin, pan Ab-5
Regulatory Status RUO
Clone ACTN05; same as C4
Isotype IgG1 kappa
Format Concentrated
Classification Monoclonal
Research Discipline Muscle Biology
Concentration 1mg/mL
Type Primary
Conjugate Unlabeled

Obtain accurate, reproducible results in immunohistochemistry, western blotting, and immunoprecipitation experiments with Thermo Scientific™ Actin, pan Ab-5, Mouse Monoclonal Antibody.

Host Species: Mouse

Clone: ACTN05; same as C4

Isotype: IgG1/κ

Species Reactivity: Human, Cow, Mouse, Dog, Pig, Rabbit, Rat, Chicken, Dictyostelium discoideum and Physarum polycephalum. Others not known.

Epitope: Not determined

Immunogen: Chicken gizzard actin

Positive Control: HeLa cells

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation (Denatured verified)

Note:

USA: RUO; Int'l: RUO