missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Pamoic acid, 99%, Alfa Aesar™

Manufacturer:  Alfa Aesar A1520730

 View more versions of this product

Catalog No. AAA1520730



Description

Description

Applications
Pamoic acid is used in preparation of repository derivatives of medicinal agents, used in pharmacology to increase the solubility of a drug in water.

Solubility
Soluble in nitrobenzene, pyridine

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

Pamoic acid
99%
MFCD00004079
14,7005
Soluble in nitrobenzene, pyridine
C1=CC=C2C(=C1)C=C(C(=C2CC3=C(C(=CC4=CC=CC=C43)C(=O)O)O)O)C(=O)O
388.375
CHEBI:50186
99%
250g
130-85-8
C23H16O6
901319
pamoic acid, embonic acid, 4,4'-methylenebis 3-hydroxy-2-naphthoic acid, pamosaeure, unii-7rrq8qz38n, 2-naphthalenecarboxylic acid, 4,4'-methylenebis 3-hydroxy, 7rrq8qz38n, pamoic acid 98+% hplc, 2-naphthalenecarboxylic acid, 4,4'-methylenebis 3-hydroxy, 4,4'-methylen-bis-3-hydroxy-2-naphthoesaeure
WLJNZVDCPSBLRP-UHFFFAOYSA-N
4-[(3-carboxy-2-hydroxynaphthalen-1-yl)methyl]-3-hydroxynaphthalene-2-carboxylic acid
8546
388.38
>300°C

Chemical Identifiers

130-85-8
388.375
WLJNZVDCPSBLRP-UHFFFAOYSA-N
8546
4-[(3-carboxy-2-hydroxynaphthalen-1-yl)methyl]-3-hydroxynaphthalene-2-carboxylic acid
C23H16O6
MFCD00004079
pamoic acid, embonic acid, 4,4'-methylenebis 3-hydroxy-2-naphthoic acid, pamosaeure, unii-7rrq8qz38n, 2-naphthalenecarboxylic acid, 4,4'-methylenebis 3-hydroxy, 7rrq8qz38n, pamoic acid 98+% hplc, 2-naphthalenecarboxylic acid, 4,4'-methylenebis 3-hydroxy, 4,4'-methylen-bis-3-hydroxy-2-naphthoesaeure
CHEBI:50186
C1=CC=C2C(=C1)C=C(C(=C2CC3=C(C(=CC4=CC=CC=C43)C(=O)O)O)O)C(=O)O
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : QL2180000

TSCA : TSCA

Documents