missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Specifications
Pamabrom | |
295°C | |
C11H18BrN5O3 | |
5 g | |
ATOTUUBRFJHZQG-UHFFFAOYSA-N | |
2-amino-2-methylpropan-1-ol;8-bromo-1,3-dimethyl-7H-purine-2,6-dione | |
11806 | |
≥98.0% (HPLC,T) |
606-04-2 | |
White | |
MFCD00867328 | |
8-Bromotheophylline 2-Amino-2-methyl-1-propanol | |
CC(C)(CO)N.CN1C2=C(C(=O)N(C1=O)C)NC(=N2)Br | |
348.201 | |
348.20 | |
Crystalline Powder |
Chemical Identifiers
606-04-2 | |
348.201 | |
ATOTUUBRFJHZQG-UHFFFAOYSA-N | |
11806 | |
CC(C)(CO)N.CN1C2=C(C(=O)N(C1=O)C)NC(=N2)Br |
C11H18BrN5O3 | |
MFCD00867328 | |
8-Bromotheophylline 2-Amino-2-methyl-1-propanol | |
2-amino-2-methylpropan-1-ol;8-bromo-1,3-dimethyl-7H-purine-2,6-dione |
Safety and Handling
TSCA : No