missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Palonosetron Hydrochloride, TCI America™

Manufacturer:  TCI America P2051200MG

 View more versions of this product

Catalog No. P2051200MG



Specifications

Specifications

Palonosetron Hydrochloride
307°C
135729-62-3
MFCD07370072
C1CC2CN(C(=O)C3=C2C(=CC=C3)C1)C4CN5CCC4CC5.Cl
332.872
CHEBI:85157
Crystal-Powder at 20°C
Undesignated
200mg
C19H25ClN2O
OLDRWYVIKMSFFB-SSPJITILSA-N
(3aS)-2-[(3S)-1-azabicyclo[2.2.2]octan-3-yl]-3a,4,5,6-tetrahydro-3H-benzo[de]isoquinolin-1-one;hydrochloride
6918303
332.87
≥98.0% (HPLC,T)

Chemical Identifiers

135729-62-3
332.872
OLDRWYVIKMSFFB-SSPJITILSA-N
CHEBI:85157
C1CC2CN(C(=O)C3=C2C(=CC=C3)C1)C4CN5CCC4CC5.Cl
C19H25ClN2O
MFCD07370072
6918303
(3aS)-2-[(3S)-1-azabicyclo[2.2.2]octan-3-yl]-3a,4,5,6-tetrahydro-3H-benzo[de]isoquinolin-1-one;hydrochloride
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

Documents