Please login to your online account to display your discounted pricing

Alfa Aesar™ Palmityltrifluoromethylketone, 98%


Manufacturer: alfa aesar™  J67117MA

 View more versions of this product

Catalog No. AAJ67117MA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Assay 98%
Assay 98%
CAS 141022-99-3
Molecular Formula C17H31F3O
Formula Weight 308.43
Synonym PACOCF3; PTK
Quantity 10mg
Description White to off-white
Storage Requirements -30°C to -10°C
Format Solid
Type Palmityltrifluoromethylketone, 98%