Please login to your online account to display your discounted pricing

Palmitoylisopropylamide, Tocris

Inhibitor of FAAH

$102.98 - $479.45

Chemical Identifiers

Chemical Name or Material Palmitoylisopropylamide
Assay Percent Range >98%
Molecular Formula C19H39NO
Formula Weight 297.52
Synonym N-(1-Methylethyl)-hexadecanamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

181510

 
r & d systems
RDS181510
10mg Each for $102.98

181550

 
r & d systems
RDS181550
50mg Each for $479.45
Description & Specifications

Specifications

Formula Weight 297.52
Quantity 10mg
Chemical Name or Material Palmitoylisopropylamide
Assay Percent Range >98%
Molecular Formula C19H39NO
CAS 189939-61-5
Synonym N-(1-Methylethyl)-hexadecanamide
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature