Please login to your online account to display your discounted pricing

Palmitoylethanolamide, Tocris


Selective GPR55 agonist; FAAH and PAA substrate

Quantity 10mg

Manufacturer: r & d systems  RDS087910

 View more versions of this product

Catalog No. 087910

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10mg
Formula Weight 299.5
Assay Percent Range >98%
Molecular Formula C18H37NO2
CAS 544-31-0
Synonym N-(2-Hydroxyethyl)hexadecanamide
Recommended Storage Store at Room Temperature
Chemical Name or Material Palmitoylethanolamide

Specifications

Chemical Name or Material Palmitoylethanolamide
Assay Percent Range >98%
Molecular Formula C18H37NO2
Formula Weight 299.5
Synonym N-(2-Hydroxyethyl)hexadecanamide
Accessories

Store at Room Temperature