Please login to your online account to display your discounted pricing

Palmitoylethanolamide, Tocris

Selective GPR55 agonist; FAAH and PAA substrate

$113.57 - $478.51

Chemical Identifiers

Chemical Name or Material Palmitoylethanolamide
Assay Percent Range >98%
Molecular Formula C18H37NO2
Formula Weight 299.5
Synonym N-(2-Hydroxyethyl)hexadecanamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

087910

 
r & d systems
RDS087910
10mg Each for $113.57

087950

 
r & d systems
RDS087950
50mg Each for $478.51
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 299.5
Assay Percent Range >98%
Molecular Formula C18H37NO2
CAS 544-31-0
Synonym N-(2-Hydroxyethyl)hexadecanamide
Recommended Storage Store at Room Temperature
Chemical Name or Material Palmitoylethanolamide
Safety and Handling

Store at Room Temperature