Learn More
Palmitoyl chloride, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 304291000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Palmitoyl chloride | |
112-67-4 | |
9.0°C to 14.0°C | |
199.0°C (2.0 mmHg) | |
Authentic | |
Glass bottle | |
1.4510 to 1.4530 | |
MFCD00000742 | |
02, 374 | |
palmitoyl chloride, palmitic acid chloride, palmitoylchloride, unii-7642r2josb, hexadecanoyl-chloride, palm acetyl chlorine 14 acetyl chlorine, hexadecanoic acid, chloride, palmitic chloride, hexadecanoylchloride, n-hexadecanoyl chloride | |
ARBOVOVUTSQWSS-UHFFFAOYSA-N | |
hexadecanoyl chloride | |
8206 | |
98% |
98% | |
1% max. (as palmitic acid) | |
0.9000g/mL | |
160°C | |
97.5% min. (GC) | |
C16H31ClO | |
CH3(CH2)14COCl | |
100 mL | |
0.9 | |
Solubility in water: decomposes. Other solubilities: soluble in ether,decomposes in alcohol | |
CCCCCCCCCCCCCCCC(=O)Cl | |
274.88 | |
274.88 | |
Liquid |
Chemical Identifiers
112-67-4 | |
ARBOVOVUTSQWSS-UHFFFAOYSA-N | |
8206 | |
CCCCCCCCCCCCCCCC(=O)Cl |
MFCD00000742 | |
palmitoyl chloride, palmitic acid chloride, palmitoylchloride, unii-7642r2josb, hexadecanoyl-chloride, palm acetyl chlorine 14 acetyl chlorine, hexadecanoic acid, chloride, palmitic chloride, hexadecanoylchloride, n-hexadecanoyl chloride | |
hexadecanoyl chloride |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
EINECSNumber : 203-996-7
TSCA : TSCA
RUO – Research Use Only