Please login to your online account to display your discounted pricing

Palmitoyl chloride, 98%, ACROS Organics™


Quantity 5mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  304290050

 View more versions of this product

Catalog No. AC304290050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay 98 to 101% (Argentometry), ≥97.5%(GC)
Physical Form Liquid
Formula Weight 274.88
Quantity 5mL
Color Clear
Beilstein 02, 374
Density 0.9000g/mL
EINECS Number 203-996-7
TSCA TSCA
Acidity 1% max. (as palmitic acid)
Name Note 98%
Infrared Spectrum Authentic
Packaging Glass bottle
Phosphorus 150ppm max.
Refractive Index 1.4510 to 1.4530 (20°C, 589nm)
Health Hazard 1 Danger
Health Hazard 2 GHS P Statement Causes severe skin burns and eye damage. Contact with water liberates toxic gas.
Health Hazard 3 GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Co
Chemical Name or Material Palmitoyl chloride
Assay Percent Range 98%
Molecular Formula C16H31ClO
Linear Formula CH3(CH2)14COCl
MDL Number MFCD00000742
CAS 112-67-4

Specifications

Chemical Name or Material Palmitoyl chloride
Name Note 98%
CAS 112-67-4
Assay 98 to 101% (Argentometry), ≥97.5%(GC)
Assay Percent Range 98%
Molecular Formula C16H31ClO
Linear Formula CH3(CH2)14COCl
Formula Weight 274.88
MDL Number MFCD00000742
Accessories

Danger

GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Co

GHS P Statement Causes severe skin burns and eye damage. Contact with water liberates toxic gas.

SDS & Safety Documents