missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Palmitic acid, sodium salt, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 416700050

 View more versions of this product

Catalog No. AC416700050



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Palmitic acid, sodium salt
White
Authentic
Glass bottle
CH3(CH2)14COONa
5 g
sodium palmitate, palmitic acid sodium salt, hexadecanoic acid, sodium salt, sodium hexadecanoate, palmitic acid, sodium salt, sodium pentadecanecarboxylate, unii-jq43kp6296, hsdb 759, hexadecanoic acid sodium salt, hexadecanoic acid,sodium salt 1:1
CCCCCCCCCCCCCCCC(=O)[O-].[Na+]
278.4
278.4
Powder
408-35-5
2% max. (110°C, 4 hrs)
98%
C16H31NaO2
MFCD00002749
02,III,94
GGXKEBACDBNFAF-UHFFFAOYSA-M
sodium;hexadecanoate
2735111
98%

Chemical Identifiers

408-35-5
278.4
GGXKEBACDBNFAF-UHFFFAOYSA-M
2735111
CCCCCCCCCCCCCCCC(=O)[O-].[Na+]
C16H31NaO2
MFCD00002749
sodium palmitate, palmitic acid sodium salt, hexadecanoic acid, sodium salt, sodium hexadecanoate, palmitic acid, sodium salt, sodium pentadecanecarboxylate, unii-jq43kp6296, hsdb 759, hexadecanoic acid sodium salt, hexadecanoic acid,sodium salt 1:1
sodium;hexadecanoate
Safety and Handling

Safety and Handling

EINECSNumber : 206-988-1

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only