missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Palmitic acid hydrazide, 99%

Manufacturer:  Alfa Aesar™ L1301306

 View more versions of this product

Catalog No. AAL1301306



Specifications

Specifications

Palmitic acid hydrazide
5g
C16H34N2O
MFCD00066357
palmitohydrazide, palmitic acid hydrazide, hexadecanoyl hydrazide, hexadecanoic acid hydrazide, palmiticacidhydrazide, hexadecanohydrazide #, acmc-1cbma, 16-hexadecanoyl hydrazide, hexadecanoic acid,hydrazide, hexdecanoic acid hydrazide
CCCCCCCCCCCCCCCC(=O)NN
270.461
270.46
150°C to 152°C
2619-88-7
CH3(CH2)14CONHNH2
1784616
SSVSELJXJJCANX-UHFFFAOYSA-N
hexadecanehydrazide
225536
99%

Chemical Identifiers

2619-88-7
270.461
SSVSELJXJJCANX-UHFFFAOYSA-N
225536
CCCCCCCCCCCCCCCC(=O)NN
C16H34N2O
MFCD00066357
palmitohydrazide, palmitic acid hydrazide, hexadecanoyl hydrazide, hexadecanoic acid hydrazide, palmiticacidhydrazide, hexadecanohydrazide #, acmc-1cbma, 16-hexadecanoyl hydrazide, hexadecanoic acid,hydrazide, hexdecanoic acid hydrazide
hexadecanehydrazide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents