missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Palmitelaidic acid, >99%, MP Biomedicals™

Supplier:  MP Biomedicals Inc 0215604491

 View more versions of this product

Catalog No. ICN15604491



Description

Description

Specifications

Specifications

Palmitoleic Acid
Colorless
≥99%
1 g
UOTMYNBWXDUBNX-UHFFFAOYSA-N
1-[(3,4-dimethoxyphenyl)methyl]-6,7-dimethoxyisoquinoline;hydrochloride
6084
>99%
61-25-6
0.895g/mL at 20°C
C20H22ClNO4
papaverine hydrochloride, cardiospan, cardoverina, dispamil, drapavel, forpavin, papalease, papaversan, pavatest, paverolan
COC1=C(C=C(C=C1)CC2=NC=CC3=CC(=C(C=C32)OC)OC)OC.Cl
375.849
254.4
Frozen Liquid

Chemical Identifiers

61-25-6
375.849
papaverine hydrochloride, cardiospan, cardoverina, dispamil, drapavel, forpavin, papalease, papaversan, pavatest, paverolan
1-[(3,4-dimethoxyphenyl)methyl]-6,7-dimethoxyisoquinoline;hydrochloride
C20H22ClNO4
UOTMYNBWXDUBNX-UHFFFAOYSA-N
6084
COC1=C(C=C(C=C1)CC2=NC=CC3=CC(=C(C=C32)OC)OC)OC.Cl
Safety and Handling

Safety and Handling

EINECSNumber : 206-765-9

Recommended Storage : Store at -0°C.

SDS
Documents

Documents

Promotions

Promotions