Please login to your online account to display your discounted pricing

Alfa Aesar™ Palladium wire, 2.0mm (0.08 in.) dia., 99.95% (metals basis)

Chemical Identifiers

Chemical Name or Material Palladium wire
CAS 7440-05-3
Name Note 2.0mm (0.08 in.) dia.
MDL Number MFCD00011167
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA14761BQ

  SDS
alfa aesar™
14761BQ
5cm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

MDL Number MFCD00011167
Weight ≈37.7g/m
CAS 7440-05-3
Assay 99.95%
Assay Percent Notes (metals basis)
Quantity 5cm
Odor Odorless
Chemical Name or Material Palladium wire
Name Note 2.0mm (0.08 in.) dia.
Physical Form Wire
TSCA TSCA